Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Isopropyl Alcohol 70%
SCI_ED

Catalog No. S25371A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25371A 4 L
S25371 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25371A
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Isopropyl alcohol solution is ideal for educational and laboratory applications.

  • Isopropyl Alcohol Solution
  • Product is for educational use.
  • Available in 500 mL or 4 L sizes.

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Density 0.785g/mL
Quantity 4 L
Assay Percent Range 70%
Linear Formula (CH3)2CHOH
Formula Weight 6.1
Chemical Name or Material Isopropyl Alcohol
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 3, II, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.