Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Isopropyl Alcohol
SCI_ED

Catalog No. S25372 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25372 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25372
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity isopropyl alcohol for cleaning, sanitizing, and lab demonstrations in educational environments.

  • Chemical Name: Isopropyl Alcohol (≥99%)
  • Formula: (CH3)2CHOH ∣ Formula Weight: 60.1 g/mol (corrected from 6.1)
  • Form: Liquid
  • Color: Colorless
  • Density: 0.785 g/mL
  • Quantity: 500 mL
  • Packaging: Lab-grade bottle
  • Ideal For: Cleaning labware, chemistry demonstrations, and classroom lab use
  • Note: For educational use, Certificate of Analysis (C of A) not provided

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Density 0.785g/mL
Quantity 500 mL
Assay Percent Range >99%
Linear Formula (CH3)2CHOH
Formula Weight 6.1
Chemical Name or Material Isopropyl Alcohol
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 3, II, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.