Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, OmniSolv™, MilliporeSigma™

Catalog No. MPX1834P1 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glas bottle (Pre packed)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX1834P1 4 L Glas bottle (Pre packed)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX1834P1 Supplier MilliporeSigma Supplier No. PX1834P1
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Acidity Titrable acid: 0.2µeq/g
Boiling Range 82.4°C (1013 hPa)
Melting Point -89.5°C
Color Colorless
Density .786 g/cc at 20°C
Flash Point 12°C
Ignition Point 425°C
Odor Alcohol-like
Packaging Glas bottle (Pre packed)
Quantity 4 L
UN Number UN 1219
Residue after Evaporation 1ppm
Vapor Pressure 43 hPa (20°C)
Formula Weight 60.10
Percent Purity 99.9%
Grade OmniSolv
Physical Form Liquid
Chemical Name or Material Isopropyl Alcohol
Show More Show Less
Hazard Category Danger
Hazard Statement Hazard Statements H225 Highly flammable liquid and vapor. H319 Causes serious eye irritation. H336 May cause drowsiness or dizziness
Precautionary Statement Precautionary Statements P210 Keep away from heat/sparks/open flames/hot surfaces. - No smoking. P233 Keep container tightly closed. P240 Ground/bond container and receiving equipment. P305 + P351 + P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P403 + P235 Store in a well-ventilated place. Keep cool
EINECSNumber 200-661-7
RTECSNumber NT8050000
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.