Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, 99.8% Minimum, Solvent Grade, Ricca Chemical
SDP

Catalog No. RSOI004055E Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
55 gal.
Packaging:
Poly Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOI004055E 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSOI004055E Supplier Ricca Chemical Company Supplier No. RSOI004055E

missing translation for 'validMainframeMsg'

Specifications

Color (APHA) Max 5
Acidity Max 0.0020
Assay Percent Range Min 99.8%
Quantity 55 gal.
Packaging Poly Drum
Specific Gravity At 20°C: 0.785-0.787
Water Max 0.10
Chemical Name or Material Isopropyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.