Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, 99.9%, MilliporeSigma™

Catalog No. 50947693 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-693 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-693 Supplier MilliporeSigma Supplier No. PX18277
Add to Cart
Edge
Add to Cart

Chemical Identifiers

CAS 67-63-0
Molecular Weight (g/mol) 60.1

Specifications

Melting Point −89.5°C
Quantity 100 mL
Flash Point 12°C
Packaging Glass Bottle
Residue after Evaporation 1 ppm max
Assay Percent Range 99.9% min
Vapor Pressure 43 hPa (20°C)
Boiling Point 82.4°C (1013 hPa)
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.