Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol 99%, StatLab™
GSA_VASDP

Catalog No. STL364201
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STL364201 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STL364201 Supplier SLMP Llc Dba Statlab Medical Supplier No. 364201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, if in contact with skin, or if inhaled.

Synonyms: Isopropanol, 2-Propanol, Propan-2-ol, Dimethylcarbinol

  • Ideal for small tissue, biopsy and microwave processing

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol

Specifications

Quantity 1 gal.
Packaging Bottle
Concentration or Composition (by Analyte or Components) 0.99
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.