Learn More
Hach Company Isopropyl Alcohol, 500 mL
Catalog No. 501995035 Shop All Hach Company Products
Encompass
Isopropanol, ACS Grade, CAS No. 67-63-0 CH3CHOHCH3, FW = 60.10 Assay: ≥99.5%
Catalog No. 50-199-5035 Supplier Hach Company Supplier No. 1445949
Isopropanol, ACS Grade, CAS No. 67-63-0 CH3CHOHCH3, FW = 60.10 Assay: ≥99.5%