Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, Exceeds A.C.S. Specifications, HPLC Grade, 99.7%, Spectrum™ Chemical
SDP

Catalog No. 18603538 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
4 L
4 x 4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-538 4 L Amber Glass Bottle
18-603-540 4 x 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-603-538 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HP6924LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

HP692, 67-63-0, C3H8O

Spectrum™ Chemical Isopropyl Alcohol, Exceeds A.C.S. Specifications, HPLC Grade solvents are lot-tested and precisely controlled to meet the high quality requirements of chromatographic analysis including high purity, low UV-absorbance, guaranteed suitability for fluorescence detection, low particulates (eliminates pre-filtration) and controlled low-content of non-volatile components. All Isopropyl Alcohol chromatography solvents are delivered with an expiration date.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100%
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 2 ppm
Water 0.05%
UN Number UN1219
Boiling Point 78.3°C
Percent Purity 99.7%
Grade High Purity
Chemical Name or Material Isopropyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.