Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol IPA 70 GMP, Greenfield

Catalog No. 502946050
Encompass
Change view
Click to view available options
Metal Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-294-6050 Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-6050 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P211000GMPDM55M
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-63-0 (Isopropyl alcohol) , 7732-18-5 (Water)
Molecular Weight (g/mol) 60.1 (Isopropyl Alcohol), 18.02 (Water)

Specifications

Quantity 55 Gallon
Packaging Metal Drum
Boiling Point 181.4 °F (83 °C)
Percent Purity 70%
Grade Reagent
Chemical Name or Material Isopropyl Alcohol 70%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.