Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol IPA 99 ACS USP and NF, Greenfield

Catalog No. 502946079
Encompass
Change view
Click to view available options
Packaging:
24x1 Poly Pint Case
4x1 Gallon Case
Metal Drum
Metal Pail
Poly Drum
Poly Pail
Shelf Sitter
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
50-294-6079 24x1 Poly Pint Case
50-294-6051 4x1 Gallon Case
50-294-6052 Shelf Sitter
50-294-6034 Poly Drum
50-294-6036 Poly Pail
50-294-6006 Metal Pail
50-294-5975 Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 50-294-6079 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P231ACS099CSPP

Please call customer service for current pricing. 1-800-766-7000

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-63-0
Molecular Weight (g/mol) 60.1

Specifications

Packaging 24x1 Poly Pint Case
Boiling Point 181.4 °F (83 °C)
Percent Purity 99%
Grade ACS/NF/USP
Chemical Name or Material Isopropyl Alcohol 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.