Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Isopropyl Alcohol, Reagent A.C.S. 1 QT
SDP

Supplier:  Electron Microscopy Sciences 16930

Encompass_Preferred

(2-Propanol)
CAS #67-63-0 CH₃CH(OH)CH₃
Boiling point: 1°C

Catalog No. 50-267-38


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.