Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, Reagent, ACS, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18603545 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-545 4 L Poly Bottle
18-603-551 125 mL Amber Glass Bottle
18-603-541 500 mL Amber Glass Bottle
18-603-543 1 L Amber Glass Bottle
18-603-542 6 x 500 mL Amber Glass Bottle
18-603-544 4 L Amber Glass Bottle
18-603-546 6 x 1 L Amber Glass Bottle
18-603-547 4 x 4 L Amber Glass Bottle
18-603-548 4 x 4 L Poly Bottle
18-603-549 20 L Steel Can
18-603-550 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18-603-545 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I10484LTPL
Only null left
Add to Cart
Add to Cart

I1048, 67-63-0, C3H8O

Spectrum™ Chemical Isopropyl Alcohol, Reagent, ACS is a compound which is colorless, flammable and has a very strong odor. Isopropyl alcohol has many uses, including as an analytical solvent reagent in the laboratory. Other chemical applications include use as a chemical synthesis intermediate, disinfectant, fuel additive and as a biological specimen preservative.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100%
Quantity 4 L
Packaging Poly Bottle
Residue after Evaporation 0.001%
UN Number UN1219
Water 0.2%
Boiling Point 78.3°C
Percent Purity 99.5%
Grade Reagent
Chemical Name or Material Isopropyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.