Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol Solution, LC-MS, For flushing LC-MS systems, 50/50 Isopropyl Alcohol / Water, Solstice

Catalog No. 6000168 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 1 L
Packaging:
Glass Bottle, Gl45
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-001-68 6 x 1 L Glass Bottle, Gl45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-001-68 Supplier Solstice Supplier No. LC7031
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-63-0 , 7732-18-5
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Boiling Point 82.3°C
CAS Max % 55.6000%, 44.4000%
UN Number UN1219
Density 0.9189 g/cm3
Grade LC-MS
Packaging Glass Bottle, Gl45
Quantity 6 x 1 L
Flash Point 22.78°C
Chemical Name or Material Isopropyl Alcohol Solution, LC-MS, For flushing LC-MS systems, 50/50 Isopropyl Alcohol / Water
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol solution
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.