Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol, Spectrophotometric Grade, ACS, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18611128 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-128 500 mL Amber Glass Bottle
18-611-129 1 L Amber Glass Bottle
18-611-130 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-611-128 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SP158500MLGL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 1
Quantity 500 mL
Packaging Amber Glass Bottle
Residue after Evaporation 1e-05
Water 0.002
Boiling Point 78.3°C
Percent Purity 0.995
Grade Spectrophotometry
Chemical Name or Material Isopropyl Alcohol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.