Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Alcohol USP, Macron Fine Chemicals™

Catalog No. MK30314 Shop All Avantor Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-3031-4 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-3031-4 Supplier Avantor Supplier No. 303108
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 67-63-0
Quantity 4 L
Packaging Glass
Molecular Formula C3H8O
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.10
PubChem CID 3776
ChEBI CHEBI:17824
Formula Weight 60.1
Grade BP/EP/FCC/JP/NF/USP
Chemical Name or Material Isopropyl Alcohol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.