Learn More
Crescent Chemical Co Inc ISOPROPYL-B-D-THIOGALACTOPYRAN
SDP
Catalog No. 50247756
Encompass_Preferred
Isopropyl-b-D-thiogalactopyranoside (IPTG) research grade, dioxane-free Analog of galactose, not split by β-galactosidase. Inducer of the lac operon in bacteria. Used in conjunction with X-Gal(5-bromo-4-chloro-indolyl-β-D-galactoside, cat. no. 15243) for detection of lac+ colonies. Dissolv
Catalog No. 50-247-756 Supplier Crescent Chemical Co Inc Supplier No. SE26600.04
Isopropyl-b-D-thiogalactopyranoside (IPTG) research grade, dioxane-free Analog of galactose, not split by β-galactosidase. Inducer of the lac operon in bacteria. Used in conjunction with X-Gal(5-bromo-4-chloro-indolyl-β-D-galactoside, cat. no. 15243) for detection of lac+ colonies. Dissolv