Learn More
Crescent Chemical Co Inc ISOPROPYL-B-D-THIOGALACTOPYRAN
SDP
Supplier:  Crescent Chemical Co Inc SE26600.06
Encompass_Preferred
Isopropyl-b-D-thiogalactopyranoside (IPTG) research grade, dioxane-free Analog of galactose, not split by β-galactosidase. Inducer of the lac operon in bacteria. Used in conjunction with X-Gal(5-bromo-4-chloro-indolyl-β-D-galactoside, cat. no. 15243) for detection of lac+ colonies. Dissolv
Catalog No. 50-247-757