Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isopropyl benzenesulfonate | 6214-18-2 | MFCD04038771 | 99.6% | 200.25 g/mol | C9H12O3S | 10 G
SDP

Catalog No. 5000210198
Encompass_Preferred

Isopropyl benzenesulfonate is a sulfonate ester provided as a research reference and described as a potential genotoxic impurity. It is a colorless to light yellow liquid with formula C9H12O3S and a molecular weight of 200.25 g/mol.

  • High reported purity: 99.61% (typical)
  • Physical state: liquid (density 1.145 g/cm³)
  • Commonly supplied in small laboratory packs, including 10 g
  • Storage: stable at room temperature for up to 3 years; in solvent store at -80°C or -20°C for extended periods
  • Intended for research use only; not for human use

Catalog No. 50-002-10198 Supplier Medchemexpress LLC Supplier No. HYW04343710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isopropyl benzenesulfonate is a sulfonate ester provided as a research reference and described as a potential genotoxic impurity. It is a colorless to light yellow liquid with formula C9H12O3S and a molecular weight of 200.25 g/mol.

  • High reported purity: 99.61% (typical)
  • Physical state: liquid (density 1.145 g/cm³)
  • Commonly supplied in small laboratory packs, including 10 g
  • Storage: stable at room temperature for up to 3 years; in solvent store at -80°C or -20°C for extended periods
  • Intended for research use only; not for human use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.