Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isopropyl benzenesulfonate | 6214-18-2 | MFCD04038771 | 99.6% | 200.25 g/mol | C9H12O3S | 100 G
SDP

Catalog No. 5000210174
Encompass_Preferred

Isopropyl benzenesulfonate is a sulfonate ester and a potential genotoxic impurity supplied for research use. It is a colorless to light yellow liquid with formula C9H12O3S and a molecular weight of 200.25 g/mol.

  • A sulfonate ester and a potential genotoxic impurity.
  • High purity: 99.6%.
  • Physical state: colorless to light yellow liquid; density 1.145 g/cm³.
  • CAS number: 6214-18-2.
  • Available package sizes: 10 g, 25 g, 50 g, 100 g, bulk on request.
  • Storage: store at room temperature for up to 3 years; in solvent store at -80°C for 2 years or -20°C for 1 year.

Catalog No. 50-002-10174 Supplier Medchemexpress LLC Supplier No. HYW043437100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isopropyl benzenesulfonate is a sulfonate ester and a potential genotoxic impurity supplied for research use. It is a colorless to light yellow liquid with formula C9H12O3S and a molecular weight of 200.25 g/mol.

  • A sulfonate ester and a potential genotoxic impurity.
  • High purity: 99.6%.
  • Physical state: colorless to light yellow liquid; density 1.145 g/cm³.
  • CAS number: 6214-18-2.
  • Available package sizes: 10 g, 25 g, 50 g, 100 g, bulk on request.
  • Storage: store at room temperature for up to 3 years; in solvent store at -80°C for 2 years or -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.