Learn More
Medchemexpress LLC Isopropyl benzoate | 939-48-0 | MFCD00048289 | 95.9% | 164.20 | C10H12O2 | 1 G
SDP
Catalog No. 5000301181
Encompass_Preferred
Isopropyl benzoate is an aromatic ester (benzoic acid isopropyl ester) used as a chemical reagent, reference standard, and fragrance/flavor ingredient. It is a colorless to light-yellow liquid (C10H12O2) with a molecular weight of 164.20 and is supplied in laboratory pack sizes from 1 g to bulk quantities.
  • Aromatic ester suitable for synthetic applications.
  • Used in fragrance and flavor formulation studies.
  • Available in small and bulk pack sizes for laboratory use.
  • Provided with purity information and supporting documentation.
  • Colorless to light-yellow liquid with reported density ~1.01 g/cm3.
Catalog No. 50-003-01181 Supplier Medchemexpress LLC Supplier No. HYW0171951G
Isopropyl benzoate is an aromatic ester (benzoic acid isopropyl ester) used as a chemical reagent, reference standard, and fragrance/flavor ingredient. It is a colorless to light-yellow liquid (C10H12O2) with a molecular weight of 164.20 and is supplied in laboratory pack sizes from 1 g to bulk quantities.
  • Aromatic ester suitable for synthetic applications.
  • Used in fragrance and flavor formulation studies.
  • Available in small and bulk pack sizes for laboratory use.
  • Provided with purity information and supporting documentation.
  • Colorless to light-yellow liquid with reported density ~1.01 g/cm3.