Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isopropyl benzoate | 939-48-0 | MFCD00048289 | 95.9% | 164.20 g/mol | C10H12O2 | 5 G
SDP

Catalog No. 5000299521
Encompass_Preferred

Isopropyl benzoate is the isopropyl ester of benzoic acid, supplied for research use as an organic reagent and reference compound. It is used in laboratory applications where an ester solvent, analytical standard, or flavor/fragrance intermediate is required.

  • Useful as an organic reagent and reference standard.
  • Functions as a solvent and flavor/fragrance intermediate.
  • Molecular formula C10H12O2 and molecular weight 164.20 g/mol.
  • CAS number 939-48-0 for unambiguous identification.
  • Available in small pack sizes suitable for laboratory use.
  • Reported purity values include 95.9% (data sheet) and 99.1% (documentation).

Catalog No. 50-002-99521 Supplier Medchemexpress LLC Supplier No. HYW0171955G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isopropyl benzoate is the isopropyl ester of benzoic acid, supplied for research use as an organic reagent and reference compound. It is used in laboratory applications where an ester solvent, analytical standard, or flavor/fragrance intermediate is required.

  • Useful as an organic reagent and reference standard.
  • Functions as a solvent and flavor/fragrance intermediate.
  • Molecular formula C10H12O2 and molecular weight 164.20 g/mol.
  • CAS number 939-48-0 for unambiguous identification.
  • Available in small pack sizes suitable for laboratory use.
  • Reported purity values include 95.9% (data sheet) and 99.1% (documentation).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.