Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isopropyl benzoate | 939-48-0 | MFCD00048289 | 95.9% | 164.20 g/mol | C10H12O2 | 50 G
SDP

Catalog No. 5000299904
Encompass_Preferred

Isopropyl benzoate is an organic ester (propan-2-yl benzoate) commonly used as a reagent, solvent, and reference standard in chemical and biochemical laboratories. Supplied as a colorless to light-yellow liquid, it is applied in synthesis, analytical assays, and formulation development where ester solvents or standards are required.

  • Used as a reagent and reference standard for synthesis and analysis.
  • Colorless to light-yellow liquid, density ~1.01 g/cm3.
  • Molecular formula C10H12O2 and molecular weight 164.20 g/mol.
  • Typical purity listed at about 95.9%; batch-specific purities may vary.
  • Available in multiple laboratory pack sizes including 50 g.
  • Documentation available: data sheet, SDS, COA, and spectral files.

Catalog No. 50-002-99904 Supplier Medchemexpress LLC Supplier No. HYW01719550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isopropyl benzoate is an organic ester (propan-2-yl benzoate) commonly used as a reagent, solvent, and reference standard in chemical and biochemical laboratories. Supplied as a colorless to light-yellow liquid, it is applied in synthesis, analytical assays, and formulation development where ester solvents or standards are required.

  • Used as a reagent and reference standard for synthesis and analysis.
  • Colorless to light-yellow liquid, density ~1.01 g/cm3.
  • Molecular formula C10H12O2 and molecular weight 164.20 g/mol.
  • Typical purity listed at about 95.9%; batch-specific purities may vary.
  • Available in multiple laboratory pack sizes including 50 g.
  • Documentation available: data sheet, SDS, COA, and spectral files.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.