Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isopropyl benzoate | 939-48-0 | MFCD00048289 | 99.1% | 164.20 g/mol | C10H12O2 | 100 G
SDP

Catalog No. 5000299767
Encompass_Preferred

Isopropyl benzoate (CAS 939-48-0) is the isopropyl ester of benzoic acid used as an organic reagent, solvent, and intermediate in research, formulation, and fragrance applications. It is supplied for laboratory use with supporting documentation (COA, SDS) and is suitable for synthesis and analytical workflows.

  • High purity 99.1%.
  • Molecular formula C10H12O2.
  • Molar mass 164.20 g/mol.
  • Available in multiple pack sizes including 100 g.
  • May cause skin sensitization; follow SDS instructions and use appropriate PPE.

Catalog No. 50-002-99767 Supplier Medchemexpress LLC Supplier No. HYW017195100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isopropyl benzoate (CAS 939-48-0) is the isopropyl ester of benzoic acid used as an organic reagent, solvent, and intermediate in research, formulation, and fragrance applications. It is supplied for laboratory use with supporting documentation (COA, SDS) and is suitable for synthesis and analytical workflows.

  • High purity 99.1%.
  • Molecular formula C10H12O2.
  • Molar mass 164.20 g/mol.
  • Available in multiple pack sizes including 100 g.
  • May cause skin sensitization; follow SDS instructions and use appropriate PPE.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.