Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Benzoate 99.0+%, TCI America™
SDP

Catalog No. B007325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B007325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B007325ML Supplier TCI America Supplier No. B007325ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 939-48-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00048289
InChI Key FEXQDZTYJVXMOS-UHFFFAOYSA-N
Synonym Benzoic Acid Isopropyl Ester
PubChem CID 13654
IUPAC Name propan-2-yl benzoate
SMILES CC(C)OC(=O)C1=CC=CC=C1

Specifications

Melting Point -26°C
Color Yellow
Boiling Point 218°C
Quantity 25 mL
Formula Weight 164.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Benzoate
EINECSNumber (3)-1356
RTECSNumber DH3150000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.