Learn More
Sigma Aldrich Fine Chemicals Biosciences Isopropyl beta D thiogalac
Catalog No. 502902555
Encompass
Isopropyl--D-thiogalactoside (IPTG) is a chemical analog of galactose that cannot be cleaved by -galactosidase. IPTG functions by binding to the lacI repressor and altering its conformation which prevents the repression of the -galactosidase coding gene lacZ.
Catalog No. 50-290-2555 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11411446001
Add to Cart
Add to Cart
Isopropyl--D-thiogalactoside (IPTG) is a chemical analog of galactose that cannot be cleaved by -galactosidase. IPTG functions by binding to the lacI repressor and altering its conformation which prevents the repression of the -galactosidase coding gene lacZ.