Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl chloroacetate, 99%

Catalog No. AAA19070AC
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA19070AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA19070AC Supplier Thermo Scientific Chemicals Supplier No. A19070AC
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 105-48-6 | C5H9ClO2 | 136.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-48-6
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00040410
InChI Key VODRWDBLLGYRJT-UHFFFAOYSA-N
Synonym isopropyl chloroacetate, chloroacetic acid isopropyl ester, acetic acid, chloro-, 1-methylethyl ester, iso-propyl chloroacetate, clch2c o och ch3 2, ccris 7748, acetic acid, chloro-, isopropyl ester, acetic acid, 2-chloro-, 1-methylethyl ester, chloroacetic acid, 1-methyl ester, monochloroacetic acid isopropyl ester
PubChem CID 7759
IUPAC Name propan-2-yl 2-chloroacetate
SMILES CC(C)OC(=O)CCl

Specifications

Density 1.08
Boiling Point 149°C to 150°C
Refractive Index 1.4205
Linear Formula ClCH2CO2CH(CH3)2
Quantity 25 mL
UN Number UN2947
Formula Weight 136.58
Percent Purity 99%
Chemical Name or Material Isopropyl chloroacetate
Hazard Category H226-H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOPROPYL CHLOROACETATE
EINECSNumber 203-301-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.