Learn More
Sigma Aldrich Isopropyl chloroformate solution
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Specific rates of solvolysis of isopropyl chloroformate in various solvents at 25c has been reported.isopropyl chloroform was used in the preparation of isopropyl cyclohexane via alkylation of cyclohexene
- It was also used in the preparation of 10-isopropyloctadecanoic acid via reaction with oleic acid in the presence of ethylaluminum sesquichloride
Specifications
Specifications
| CAS | 108-23-6 |
| Density | 0.892 g/mL (at 25°C) |
| Quantity | 800 mL |
| Linear Formula | (CH3)2 CHOCOCl |
| MDL Number | MFCD00075068 |
| Molecular Weight (g/mol) | 122.55 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.