Learn More
Cayman Chemical Isopropyl dodec-11-enylfluoro
SDP
Catalog No. 502952628
Encompass_Preferred
An organophosphorus ester that antagonizes the CB1 receptor and inhibits FAAH with similar potencies (IC50s = 2 nM)
Catalog No. 50-295-2628 Supplier Cayman Chemical Supplier No. 116131 MG
An organophosphorus ester that antagonizes the CB1 receptor and inhibits FAAH with similar potencies (IC50s = 2 nM)