Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isopropyl ferulate | 59831-94-6 | 5 MG
SDP

Catalog No. 5000418843
Encompass_Preferred

Isopropyl ferulate, isolated from Notopterygium incisum, is utilized in the reduction of pharmaceuticals, preparation of antifungal agents, cosmetics, and as an antioxidant agent.

  • Isolated from Notopterygium incisum
  • Utilized in pharmaceutical reduction
  • Used in antifungal agent preparation
  • Suitable for cosmetic applications
  • Functions as an antioxidant agent
  • Purity of 99.97%
  • Molecular weight of 236.26
  • Chemical formula C13H16O4
  • Appears as a white to off-white solid
  • Classified as a phenylpropanoid and monophenol

Catalog No. 50-004-18843 Supplier Medchemexpress LLC Supplier No. HYN42035MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isopropyl ferulate, isolated from Notopterygium incisum, is utilized in the reduction of pharmaceuticals, preparation of antifungal agents, cosmetics, and as an antioxidant agent.

  • Isolated from Notopterygium incisum
  • Utilized in pharmaceutical reduction
  • Used in antifungal agent preparation
  • Suitable for cosmetic applications
  • Functions as an antioxidant agent
  • Purity of 99.97%
  • Molecular weight of 236.26
  • Chemical formula C13H16O4
  • Appears as a white to off-white solid
  • Classified as a phenylpropanoid and monophenol

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.