Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Formate, MP Biomedicals

Catalog No. ICN202538 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202538 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202538 Supplier MP Biomedicals, Inc Supplier No. 0520253825

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 625-55-8
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00014127
InChI Key RMOUBSOVHSONPZ-UHFFFAOYSA-N
Synonym isopropyl formate, formic acid, 1-methylethyl ester, formic acid isopropyl ester, formic acid, isopropyl ester, isopropyl methanoate, 1-methylethyl formate, methylethyl formate, unii-h1l164w42g, fema no. 2944, hcooch ch3 2
PubChem CID 12257
IUPAC Name propan-2-yl formate
SMILES CC(C)OC=O

Specifications

Quantity 100 g
Formula Weight 88.11
Chemical Name or Material Isopropyl Formate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.