Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isopropyl isocyanate

Catalog No. 14107025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-107-025G 25 g
14-107-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-107-025G Supplier Sigma Aldrich Supplier No. 14107025G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Isopropyl isocyanate was used as reagent during the synthesis of o-aryl n-isopropylcarbamates
  • It was used as derivatization reagent during the stereoisomeric analysis of secondary alcohols

Specifications

CAS 1795-48-8
Density 0.866 g/mL (at 25°C (literature))
Boiling Point 74°C to 75°C (lit.)
Molecular Formula C4H7NO
Refractive Index n20/D 1.382 (literature)
Linear Formula (CH3)2 CHNCO
MDL Number MFCD00002037
Quantity 25 g
Molecular Weight (g/mol) 85.1
Percent Purity ≥98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.