Learn More
Medchemexpress LLC Isopropyl myristate | 110-27-0 | 98.0% | 270.45 g/mol | C17H34O2 | 50G
SDP
Catalog No. 5000237007
Encompass_Preferred
Isopropyl myristate is the ester of isopropyl alcohol and myristic acid, used as a polar emollient and solvent in topical and cosmetic formulations to improve skin absorption and act as a vehicle for transdermal delivery. This high-purity material is suitable for formulation development and laboratory applications.
  • Acts as a polar emollient to improve skin feel and spreadability.
  • Enhances transdermal penetration of active ingredients.
  • Low-viscosity solvent compatible with oils and topical bases.
  • High purity suitable for formulation and analytical work.
  • Stable liquid at room temperature with long shelf life.
Catalog No. 50-002-37007 Supplier Medchemexpress LLC Supplier No. HY12419050G
Isopropyl myristate is the ester of isopropyl alcohol and myristic acid, used as a polar emollient and solvent in topical and cosmetic formulations to improve skin absorption and act as a vehicle for transdermal delivery. This high-purity material is suitable for formulation development and laboratory applications.
  • Acts as a polar emollient to improve skin feel and spreadability.
  • Enhances transdermal penetration of active ingredients.
  • Low-viscosity solvent compatible with oils and topical bases.
  • High purity suitable for formulation and analytical work.
  • Stable liquid at room temperature with long shelf life.