Learn More
Medchemexpress LLC Isopropyl myristate | 110-27-0 | MFCD00008982 | 98.0% | 270.45 g/mol | C17H34O2 | 100G
SDP
Catalog No. 5000237996
Encompass_Preferred
Isopropyl myristate is the isopropyl ester of myristic acid, supplied as a colorless to light yellow liquid for laboratory research. It acts as a polar emollient and solvent to improve skin absorption and to enhance transdermal delivery of compounds. This product is for research use only and is not sold to patients.
  • Acts as a polar emollient for topical formulations.
  • Enhances transdermal delivery and skin absorption.
  • High purity suitable for research applications (98.0%).
  • Colorless to light yellow liquid with density approximately 0.85 g/cm³.
  • Available in multiple pack sizes including 100 g for bulk use.
Catalog No. 50-002-37996 Supplier Medchemexpress LLC Supplier No. HY124190100G
Isopropyl myristate is the isopropyl ester of myristic acid, supplied as a colorless to light yellow liquid for laboratory research. It acts as a polar emollient and solvent to improve skin absorption and to enhance transdermal delivery of compounds. This product is for research use only and is not sold to patients.
  • Acts as a polar emollient for topical formulations.
  • Enhances transdermal delivery and skin absorption.
  • High purity suitable for research applications (98.0%).
  • Colorless to light yellow liquid with density approximately 0.85 g/cm³.
  • Available in multiple pack sizes including 100 g for bulk use.