Learn More
Medchemexpress LLC Isopropyl myristate | 110-27-0 | MFCD00008982 | 98.0% | 270.45 g/mol | C17H34O2 | 10MM 1ML
SDP
Catalog No. 5000236086
Encompass_Preferred
Isopropyl myristate is a fatty acid ester used as an emollient, solvent, and penetration enhancer in topical and formulation research. Supplied in a 10 mM solution in DMSO (1 mL vial) and available as a neat liquid in larger sizes, it is commonly used for formulation screening, solvent compatibility testing, and skin absorption studies.
Catalog No. 50-002-36086 Supplier Medchemexpress LLC Supplier No. HY12419010MM 1ML
Isopropyl myristate is a fatty acid ester used as an emollient, solvent, and penetration enhancer in topical and formulation research. Supplied in a 10 mM solution in DMSO (1 mL vial) and available as a neat liquid in larger sizes, it is commonly used for formulation screening, solvent compatibility testing, and skin absorption studies.