Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isopropyl myristate | 110-27-0 | MFCD00008982 | 98.0% | 270.45 g/mol | C17H34O2 | 25G
SDP

Catalog No. 5000238444
Encompass_Preferred

Isopropyl myristate is a fatty acid ester used as a solvent, emollient, and penetration enhancer in formulation and laboratory applications. The reagent is supplied as a liquid with typical purity 98.0% and is suitable as an excipient, analytical standard, or solvent for biochemical assays and formulation research.

  • Purity 98.0%.
  • Appearance colorless to light yellow liquid.
  • Chemical formula C17H34O2; molecular weight 270.45 g/mol.
  • Density 0.85 g/cm3.
  • Solubility in DMSO 100 mg/mL; ultrasonic aid recommended.
  • Storage (pure) -20°C for 3 years or 4°C for 2 years.

Catalog No. 50-002-38444 Supplier Medchemexpress LLC Supplier No. HY12419025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Isopropyl myristate is a fatty acid ester used as a solvent, emollient, and penetration enhancer in formulation and laboratory applications. The reagent is supplied as a liquid with typical purity 98.0% and is suitable as an excipient, analytical standard, or solvent for biochemical assays and formulation research.

  • Purity 98.0%.
  • Appearance colorless to light yellow liquid.
  • Chemical formula C17H34O2; molecular weight 270.45 g/mol.
  • Density 0.85 g/cm3.
  • Solubility in DMSO 100 mg/mL; ultrasonic aid recommended.
  • Storage (pure) -20°C for 3 years or 4°C for 2 years.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.