Learn More
Selleck Chemical LLC Isopropyl myristate S2428-25mg
SDP
Catalog No. 503112142
Encompass_Preferred
Isopropyl myristate (IPM) is used as a substitute for natural oils in cosmetics and a co-solvent with skin penetration enhancement properties of active ingredients in many topical and transdermal preparations It affects the lamellar lipid assembly in terms of bilayer perturbation and disordering
Catalog No. 50-311-2142 Supplier Selleck Chemical LLC Supplier No. S242825MG
Isopropyl myristate (IPM) is used as a substitute for natural oils in cosmetics and a co-solvent with skin penetration enhancement properties of active ingredients in many topical and transdermal preparations It affects the lamellar lipid assembly in terms of bilayer perturbation and disordering