Learn More
Sigma Aldrich Fine Chemicals Biosciences Isopropyl myristate 98
Catalog No. 502902243
Encompass
Isopropyl myristate is mainly used as a solubilizer emulsifier and emollient in cosmetic and topical medicines. It is also used as a flavoring agent in the food industry.
Catalog No. 50-290-2243 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W3556908KGK
Add to Cart
Add to Cart
Isopropyl myristate is mainly used as a solubilizer emulsifier and emollient in cosmetic and topical medicines. It is also used as a flavoring agent in the food industry.