Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Myristate, for sterility testing of ophthalmic ointments, Solstice

Catalog No. 6002154 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-021-54 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-021-54 Supplier Solstice Supplier No. 2254
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-27-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
InChI Key AXISYYRBXTVTFY-UHFFFAOYSA-N
Synonym isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester
PubChem CID 8042
IUPAC Name propan-2-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC(C)C

Specifications

CAS Max % 100.0000%
Density 0.853 g/cm3
Boiling Point 193°C
Flash Point 164°C
Packaging Glass Bottle
Quantity 4 x 4 L
Formula Weight 270.45
Chemical Name or Material Isopropyl Myristate, B&J Brand™, for sterility testing of ophthalmic ointments
ShelfLife 547 days from date of manufacture
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.