Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl n-Octanoate 97.0+%, TCI America™
SDP

Catalog No. O015725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O015725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O015725ML Supplier TCI America Supplier No. O015725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5458-59-3
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.295
MDL Number MFCD00059256
InChI Key WCGIIHOFOFCKSM-UHFFFAOYSA-N
Synonym n-Octanoic Acid Isopropyl Ester
PubChem CID 21606
IUPAC Name propan-2-yl octanoate
SMILES CCCCCCCC(=O)OC(C)C

Specifications

Color Colorless
Boiling Point 125°C
Quantity 25 mL
Formula Weight 186.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl n-Octanoate
EINECSNumber (2)-0782
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.