Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Nitrate 99%, Thermo Scientific™

Catalog No. AC412860250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC412860250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC412860250 Supplier Thermo Scientific Chemicals Supplier No. 412860250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1712-64-7
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
InChI Key GAPFWGOSHOCNBM-UHFFFAOYSA-N
Synonym isopropyl nitrate, nitric acid, 1-methylethyl ester, 2-propyl nitrate, nitric acid, isopropyl ester, isopropylnitrate, isopropylester kyseliny dusicne, isopropylester kyseliny dusicne czech, ch3 2chono2, isonite, iso-propylnitrate
PubChem CID 15575
IUPAC Name propan-2-yl nitrate
SMILES CC(C)O[N+](=O)[O-]

Specifications

Color Colorless
Density 1.03
Infrared Spectrum Authentic
Refractive Index 1.3900 to 1.3920 (20°C, 589nm)
Linear Formula (CH3)2CHONO2
Quantity 25 g
Beilstein 01, 363
Formula Weight 105.09
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Isopropyl nitrate, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.