Learn More
Medchemexpress LLC Isopropyl palmitate | 142-91-6 | 99.23% | 298.51 | 100 G
SDP
Catalog No. 5000455018
Encompass_Preferred
Isopropyl palmitate is a fatty acid ester used for the design and characterization of bioactive bilayer films. These bilayer membranes can scavenge free radicals, inhibit lipid peroxidation, and inhibit the growth of foodborne pathogens. It can also serve as an excipient, functioning as a lubricant, oily carrier, solvent, or in controlled-release transdermal films. Pharmaceutical excipients are inactive ingredients that enhance stability, solubility, and processability of pharmaceutical preparations, and can influence the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Used as the oil phase in microemulsion systems for cosmetics and drug delivery.
  • Acts as a skin penetration enhancer, promoting drug permeability and improving the efficacy of transdermal drug delivery systems.
Catalog No. 50-004-55018 Supplier Medchemexpress LLC Supplier No. HYW011426100G
Isopropyl palmitate is a fatty acid ester used for the design and characterization of bioactive bilayer films. These bilayer membranes can scavenge free radicals, inhibit lipid peroxidation, and inhibit the growth of foodborne pathogens. It can also serve as an excipient, functioning as a lubricant, oily carrier, solvent, or in controlled-release transdermal films. Pharmaceutical excipients are inactive ingredients that enhance stability, solubility, and processability of pharmaceutical preparations, and can influence the absorption, distribution, metabolism, and elimination (ADME) processes of co-administered drugs.
  • Used as the oil phase in microemulsion systems for cosmetics and drug delivery.
  • Acts as a skin penetration enhancer, promoting drug permeability and improving the efficacy of transdermal drug delivery systems.