Learn More
Medchemexpress LLC Isopropyl ((R)-(perfluorophenoxy)(phenoxy)phosphoryl)-L-alaninate | 1337529-56-2 | 96.98% | C18H17F5NO5P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY59121A5MG
Encompass_Preferred
Isopropyl ((R)-(perfluorophenoxy)(phenoxy)phosphoryl)-L-alaninate is a compound used to synthesize Sofosbuvir, helping to prevent the formation of sofosbuvir degradation impurities. It is stable in powder form for extended periods and in solvent under appropriate conditions.
  • Used in the synthesis of Sofosbuvir
  • Helps avoid degradation impurities
  • Stable in powder form for up to 3 years at -20°C and 2 years at 4°C
  • Stable in solvent at -80°C for up to 6 months and 1 month at -20°C
Catalog No. 50-003-59051