Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Isopropyl salicylate, 99%

Click to view available options
Quantity:
25g
100g
607-85-2
C10H12O3
180.203
MFCD00035703
YEULQIJMIOWCHB-UHFFFAOYSA-N
isopropyl salicylate,isopropyl 2-hydroxybenzoate,salicylic acid, isopropyl ester,iso-propyl salicylate,salicylic acid isopropyl ester,isopropyl o-hydroxybenzoate,benzoic acid, 2-hydroxy-, 1-methylethyl ester,1-methylethyl 2-hydroxybenzoate,methylethyl 2-hydroxybenzoate,opropyl salicylate
11838
CHEBI:38703
propan-2-yl 2-hydroxybenzoate
CC(C)OC(=O)C1=CC=CC=C1O
607-85-2
C10H12O3
180.203
MFCD00035703
YEULQIJMIOWCHB-UHFFFAOYSA-N
isopropyl salicylate,isopropyl 2-hydroxybenzoate,salicylic acid, isopropyl ester,iso-propyl salicylate,salicylic acid isopropyl ester,isopropyl o-hydroxybenzoate,benzoic acid, 2-hydroxy-, 1-methylethyl ester,1-methylethyl 2-hydroxybenzoate,methylethyl 2-hydroxybenzoate,opropyl salicylate
11838
CHEBI:38703
propan-2-yl 2-hydroxybenzoate
CC(C)OC(=O)C1=CC=CC=C1O
Flash Point >93°C (199°F)
Boiling Point 122°C (18mmHg)
Quantity 25g
Formula Weight 180.2
Percent Purity 99%
Chemical Name or Material Isopropyl salicylate