Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl salicylate, 99%

Catalog No. AC412892500
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412892500 250 g Glass bottle
AC412891000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC412892500 Supplier Thermo Scientific Chemicals Supplier No. 412892500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Isopropyl salicylate, 99%, Quantity: 250g, Packaging: Glass bottle, Boiling Point: 122.0 deg.C (18.0 mmHg), Colorless to Yellow, Molecular Weight: 180.203, Percent Purity: 99%, Assay Percent Range: 99%, CAS: 607-85-2, ChEBI: CHEBI:38703, Density: 1.0600g/mL, EINECS Number: 210-143-2 | CAS: 607-85-2 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-85-2
MDL Number MFCD00035703
InChI Key YEULQIJMIOWCHB-UHFFFAOYSA-N
Synonym isopropyl salicylate, isopropyl 2-hydroxybenzoate, salicylic acid, isopropyl ester, iso-propyl salicylate, salicylic acid isopropyl ester, isopropyl o-hydroxybenzoate, benzoic acid, 2-hydroxy-, 1-methylethyl ester, 1-methylethyl 2-hydroxybenzoate, methylethyl 2-hydroxybenzoate, opropyl salicylate
PubChem CID 11838
ChEBI CHEBI:38703
IUPAC Name propan-2-yl 2-hydroxybenzoate
SMILES CC(C)OC(=O)C1=CC=CC=C1O

Specifications

Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 122.0°C (18.0 mmHg)
Flash Point >93°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C10H12O3
Refractive Index 1.5090 to 1.5110
Quantity 250 g
Specific Gravity 1.06
Molecular Weight (g/mol) 180.203
Formula Weight 180.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isopropyl salicylate, 99%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-143-2
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.