Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Salicylate 99.0+%, TCI America™
SDP

Catalog No. S001425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001425ML Supplier TCI America Supplier No. S001425ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 607-85-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00035703
InChI Key YEULQIJMIOWCHB-UHFFFAOYSA-N
Synonym isopropyl salicylate, isopropyl 2-hydroxybenzoate, salicylic acid, isopropyl ester, iso-propyl salicylate, salicylic acid isopropyl ester, isopropyl o-hydroxybenzoate, benzoic acid, 2-hydroxy-, 1-methylethyl ester, 1-methylethyl 2-hydroxybenzoate, methylethyl 2-hydroxybenzoate, opropyl salicylate
PubChem CID 11838
ChEBI CHEBI:38703
IUPAC Name propan-2-yl 2-hydroxybenzoate
SMILES CC(C)OC(=O)C1=CC=CC=C1O

Specifications

Color Yellow
Boiling Point 235°C
Quantity 25 mL
Formula Weight 180.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Salicylate
EINECSNumber (3)-1585
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.