Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isopropyl stearate | 112-10-7 | MFCD00026666 | 98.0% | 326.56 g·mol⁻¹ | C21H42O2 | 250 MG
SDP

Catalog No. 5000281518
Encompass_Preferred

Isopropyl stearate (stearic acid isopropyl ester) is a fatty acid ester used as an emollient, solvent, and reagent in formulation and laboratory research. It is supplied at high purity for analytical and formulation work, with safety and quality documents available and recommended storage conditions for solid and solution forms.

  • Used as an emollient, solvent, and reagent.
  • High purity: 98.0%.
  • Molecular formula C21H42O2; molecular weight 326.56 g·mol⁻¹.
  • SDS and COA available for handling and quality assurance.
  • Store at room temperature for solids; in solvent, store at -20°C to -80°C.

Catalog No. 50-002-81518 Supplier Medchemexpress LLC Supplier No. HYW415007250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isopropyl stearate (stearic acid isopropyl ester) is a fatty acid ester used as an emollient, solvent, and reagent in formulation and laboratory research. It is supplied at high purity for analytical and formulation work, with safety and quality documents available and recommended storage conditions for solid and solution forms.

  • Used as an emollient, solvent, and reagent.
  • High purity: 98.0%.
  • Molecular formula C21H42O2; molecular weight 326.56 g·mol⁻¹.
  • SDS and COA available for handling and quality assurance.
  • Store at room temperature for solids; in solvent, store at -20°C to -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.