Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isopropylamine

Catalog No. 47129125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
250 mL
1 L
2 L
18 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-129-125ML 25 mL
47-129-118L 18 L
47-129-12L 2 L
47-129-11L 1 L
47-129-1250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 47-129-125ML Supplier Sigma Aldrich Supplier No. 47129125ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Isopropylamine (i-prnh2), a primary mine, is a urinary metabolite of propranolol.isopropylamine was used in the following studies:
  • Preparation of enantiomerically pure (r and s)- acebutolol
  • As template during the hydrothermal synthesis of sapo materials
  • Synthesis of bis(salicylaldiminate)nickel(ii) complexes
  • Preparation of hydrated, microporous aluminum phosphate alpo-14

Specifications

CAS 75-31-0
Density 0.688 g/mL (at 20°C (literature))
Boiling Point 33°C to 34°C (lit.)
Molecular Formula C3H9N
Refractive Index n20/D 1.374 (literature)
Linear Formula (CH3)2 CHNH2
MDL Number MFCD00008082
Quantity 25 mL
Synonym 2-Aminopropane
Molecular Weight (g/mol) 59.11
Percent Purity ≥99.5%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.