Learn More
Medchemexpress LLC α-isopropylaminopentiophenone hydrochloride | 18268-14-9 | 100.0% | 255.78 | C14H22ClNO | 1 ML
SDP
Catalog No. 5000229615
Encompass_Preferred
α-Isopropylaminopentiophenone hydrochloride is an α-aminoketone derivative supplied for research use. It is available as a 10 mM solution (1 mL in DMSO) or as solid milligram quantities. CAS 18268-14-9; molecular formula C14H22ClNO; molecular weight 255.78.
  • Hydrochloride salt form for improved solubility and stability.
  • Available as 10 mM solution in DMSO (1 mL) and as solid quantities.
  • High purity (99.98% assay) suitable for biochemical research.
  • Manufacturer-provided solubility and in vivo formulation recommendations.
  • Stable when stored sealed and away from moisture; solvent storage: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-29615 Supplier Medchemexpress LLC Supplier No. HY11871010MM 1ML
α-Isopropylaminopentiophenone hydrochloride is an α-aminoketone derivative supplied for research use. It is available as a 10 mM solution (1 mL in DMSO) or as solid milligram quantities. CAS 18268-14-9; molecular formula C14H22ClNO; molecular weight 255.78.
  • Hydrochloride salt form for improved solubility and stability.
  • Available as 10 mM solution in DMSO (1 mL) and as solid quantities.
  • High purity (99.98% assay) suitable for biochemical research.
  • Manufacturer-provided solubility and in vivo formulation recommendations.
  • Stable when stored sealed and away from moisture; solvent storage: -80°C (6 months), -20°C (1 month).