Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. I07875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07875G 5 g
I0787-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07875G Supplier TCI America Supplier No. I07875G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 80041-89-0
Molecular Formula C3H9BO2
MDL Number MFCD01319021
InChI Key QIPHSSYCQCBJAX-UHFFFAOYSA-N
PubChem CID 2734750

Specifications

Melting Point 115°C
Color White
Quantity 5 g
Synonym isopropylboronic acid, propan-2-yl boronic acid, 2-propaneboronic acid, prop-2-yl boronic acid, 1-methylethyl boronic acid, boronic acid, 1-methylethyl, isopropylboric acid, pubchem7987, isopropyl boronic acid, acmc-209piu
SMILES B(C(C)C)(O)O
IUPAC Name propan-2-ylboronic acid
Molecular Weight (g/mol) 87.913
Formula Weight 87.91
Physical Form Crystalline Powder
Chemical Name or Material Isopropylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.