Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isopropylmagnesium Bromide Solution

Catalog No. 775797800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
2 g
100 mL
4 x 25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-579-7800ML 4 x 25 mL
77-579-7 100 mL
77-579-7100ML 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 77-579-7800ML Supplier Sigma Aldrich Supplier No. 775797800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 920-39-8
Density 0.982 g/mL at 25°C
Molecular Formula C3H7BrMg
Linear Formula (CH3)2MgBr
MDL Number MFCD00672110
Quantity 4 x 25 mL
Molecular Weight (g/mol) 147.3 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.